CH4026586574 ChemBridge Corporation

1-(3-chloro-4-methoxyphenyl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD03044701
  • Cas Number : 63674-98-6
  • Molecular Weight : 269.681

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6760811

Safety Information

MSDS https://www.hit2lead.com/sds/6760811_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart