CH4026586579 ChemBridge Corporation

ethyl 2-chloro-5-(5-formyl-2-furyl)benzoate

  • MDL number MFCD01158422
  • Cas Number : 752231-43-9
  • Molecular Weight : 278.6877

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6782256

Safety Information

MSDS https://www.hit2lead.com/sds/6782256_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart