CH4026586581 ChemBridge Corporation

(4-ethoxyphenoxy)acetic acid

  • MDL number MFCD03419318
  • Cas Number : 5327-91-3
  • Molecular Weight : 196.1999

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6792402

Safety Information

MSDS https://www.hit2lead.com/sds/6792402_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart