CH4026586582 ChemBridge Corporation

3-methyl-2-(2-methylphenoxy)butanoic acid

  • MDL number MFCD05863708
  • Cas Number : 63403-13-4
  • Molecular Weight : 208.2536

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6800376

Safety Information

MSDS https://www.hit2lead.com/sds/6800376_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart