CH4026586591 ChemBridge Corporation

3-[(4-amino-6-oxo-1,6-dihydropyrimidin-2-yl)thio]propanoic acid

  • MDL number MFCD20503013
  • Cas Number : 63916-09-6
  • Molecular Weight : 215.2297

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6873428

Safety Information

MSDS https://www.hit2lead.com/sds/6873428_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart