CH4026586601 ChemBridge Corporation

2-[(6-chloro-4-ethyl-2-oxo-2H-chromen-7-yl)oxy]propanoic acid

  • MDL number MFCD03848270
  • Cas Number : 840479-48-3
  • Molecular Weight : 296.703

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7102136

Safety Information

MSDS https://www.hit2lead.com/sds/7102136_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart