CH4026586606 ChemBridge Corporation

2-[(6-chloro-3,4-dimethyl-2-oxo-2H-chromen-7-yl)oxy]propanoic acid

  • MDL number MFCD05863721
  • Cas Number : 853892-42-9
  • Molecular Weight : 296.703

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7109322

Safety Information

MSDS https://www.hit2lead.com/sds/7109322_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart