CH4026586614 ChemBridge Corporation

1-(4-chlorobenzoyl)piperidine-4-carboxylic acid

  • MDL number MFCD03014156
  • Cas Number : 379724-54-6
  • Molecular Weight : 267.7082

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7361053

Safety Information

MSDS https://www.hit2lead.com/sds/7361053_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart