CH4040916319 ChemBridge Corporation

2-[(2-methylbutanoyl)amino]benzoic acid

  • MDL number MFCD03899923
  • Cas Number : 713493-20-0
  • Molecular Weight : 221.2524

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7925510

Safety Information

MSDS https://www.hit2lead.com/sds/7925510_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart