CH4040922578 ChemBridge Corporation

5-oxo-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinoline-7-carbaldehyde

  • MDL number MFCD03198261
  • Cas Number : 111724-62-0
  • Molecular Weight : 213.2319

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7953410

Safety Information

MSDS https://www.hit2lead.com/sds/7953410_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart