CH4040923633 ChemBridge Corporation

4-amino-N-(1-phenylethyl)benzamide

  • MDL number MFCD01666807
  • Cas Number : 85592-75-2
  • Molecular Weight : 240.3003

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7965646

Safety Information

MSDS https://www.hit2lead.com/building-blocks/7965646
Availability details for
Quantity
  • Availability
  • Add to Cart