CH4046993967 ChemBridge Corporation

3-[(4,6-dimethyl-2-pyrimidinyl)amino]phenol

  • MDL number MFCD01872025
  • Cas Number : 81261-83-8
  • Molecular Weight : 215.2511

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7928507

Safety Information

MSDS https://www.hit2lead.com/sds/7928507_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart