CH4048389057 ChemBridge Corporation

2-amino-N-[2-(1-cyclohexen-1-yl)ethyl]benzamide

  • MDL number MFCD06624317
  • Cas Number : 825657-70-3
  • Molecular Weight : 244.3321

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7941081

Safety Information

MSDS https://www.hit2lead.com/sds/7941081_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart