CH4048389255 ChemBridge Corporation

3-(3-amino-5-oxo-4,5-dihydro-1,2,4-triazin-6-yl)propanoic acid

  • MDL number MFCD00707817
  • Cas Number : 220459-24-5
  • Molecular Weight : 184.1527

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5571200

Safety Information

MSDS https://www.hit2lead.com/sds/5571200_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart