CH4049285644 ChemBridge Corporation

4-[3-(2-chlorophenyl)-1,2,4-oxadiazol-5-yl]butanoic acid

  • MDL number MFCD06624152
  • Cas Number : 827014-22-2
  • Molecular Weight : 266.6803

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7939291

Safety Information

MSDS https://www.hit2lead.com/sds/7939291_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart