CH4061491418 ChemBridge Corporation

3-amino-N-(4-methylphenyl)benzamide

  • MDL number MFCD00443925
  • Cas Number : 14315-26-5
  • Molecular Weight : 226.2738

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5249590

Safety Information

MSDS https://www.hit2lead.com/sds/5249590_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart