CH4061491794 ChemBridge Corporation

4-(2-furoylamino)-3-methylbenzoic acid

  • MDL number MFCD07357963
  • Cas Number : 435288-00-9
  • Molecular Weight : 245.2307

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7929674

Safety Information

MSDS https://www.hit2lead.com/sds/7929674_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart