CH4061497357 ChemBridge Corporation

4-(allyloxy)-3-ethoxybenzoic acid

  • MDL number MFCD06626107
  • Cas Number : 831198-97-1
  • Molecular Weight : 222.2372

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7956005

Safety Information

MSDS https://www.hit2lead.com/sds/7956005_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart