CH4061519578 ChemBridge Corporation

2-{[(2-methoxyethyl)amino]carbonyl}cyclohexanecarboxylic acid

  • MDL number MFCD05128332
  • Cas Number : 817632-37-4
  • Molecular Weight : 229.2729

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7990689

Safety Information

MSDS https://www.hit2lead.com/building-blocks/7990689
Availability details for
Quantity
  • Availability
  • Add to Cart