CH4061520507 ChemBridge Corporation

2-(2-oxobenzo[cd]indol-1(2H)-yl)propanoic acid

  • MDL number MFCD06633017
  • Cas Number : 838850-72-9
  • Molecular Weight : 241.242

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7991949

Safety Information

MSDS https://www.hit2lead.com/sds/7991949_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart