CH4061524953 ChemBridge Corporation

1-(4-bromobenzyl)-1H-imidazole

  • MDL number MFCD06421442
  • Cas Number : 72459-46-2
  • Molecular Weight : 237.0959

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9001713

Safety Information

MSDS https://www.hit2lead.com/building-blocks/9001713
Availability details for
Quantity
  • Availability
  • Add to Cart