CH4061526174 ChemBridge Corporation

4-(2-aminobenzoyl)-2-piperazinone

  • MDL number MFCD05666702
  • Cas Number : 671794-74-4
  • Molecular Weight : 219.2398

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9002402

Safety Information

MSDS https://www.hit2lead.com/building-blocks/9002402
Availability details for
Quantity
  • Availability
  • Add to Cart