CH4061528641 ChemBridge Corporation

2-(benzylthio)-4-hydroxy-5-pyrimidinecarboxylic acid

  • MDL number MFCD02675547
  • Cas Number : 93185-33-2
  • Molecular Weight : 262.2844

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9007270

Safety Information

MSDS https://www.hit2lead.com/sds/9007270_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart