CH4064014738 ChemBridge Corporation

4-[2-(4-quinolinyl)vinyl]phenol

  • MDL number MFCD07359593
  • Cas Number : 1902965-41-6
  • Molecular Weight : 247.2912

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7992195

Safety Information

MSDS https://www.hit2lead.com/sds/7992195_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart