CH4064741192 ChemBridge Corporation

[1-(4-fluorobenzyl)-3-oxo-2-piperazinyl]acetic acid

  • MDL number MFCD05666726
  • Cas Number : 1033600-10-0
  • Molecular Weight : 266.2682

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9020055

Safety Information

MSDS https://www.hit2lead.com/sds/9020055_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart