CH4064741354 ChemBridge Corporation

[1-(3-methylbenzyl)-3-oxo-2-piperazinyl]acetic acid

  • MDL number MFCD05870475
  • Cas Number : 1023919-68-7
  • Molecular Weight : 262.3043

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9031832

Safety Information

MSDS https://www.hit2lead.com/building-blocks/9031832
Availability details for
Quantity
  • Availability
  • Add to Cart