CH4064741392 ChemBridge Corporation

(5-bromo-2-methoxybenzyl)methylamine

  • MDL number MFCD02040868
  • Cas Number : 137469-70-6
  • Molecular Weight : 230.1017

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5539963

Safety Information

MSDS https://www.hit2lead.com/sds/5539963_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart