CH4065294315 ChemBridge Corporation

2-(1H-imidazol-1-yl)-1-phenylethanone

  • MDL number MFCD00107975
  • Cas Number : 24155-34-8
  • Molecular Weight : 186.2099

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5324110

Safety Information

MSDS https://www.hit2lead.com/sds/5324110_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart