CH4065294339 ChemBridge Corporation

2-(2-fluorophenoxy)-N-(2-methoxyethyl)ethanamine

  • MDL number MFCD02633061
  • Cas Number : 420102-44-9
  • Molecular Weight : 213.2486

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5738005

Safety Information

MSDS https://www.hit2lead.com/sds/5738005_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart