CH4066472183 ChemBridge Corporation

3-(2-furoylamino)-2-methylbenzoic acid

  • MDL number MFCD07113719
  • Cas Number : 878426-41-6
  • Molecular Weight : 245.2307

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9018998

Safety Information

MSDS https://www.hit2lead.com/sds/9018998_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart