CH4068057339 ChemBridge Corporation

2-methyl-3-[(2-pyridinylmethyl)amino]benzoic acid

  • MDL number MFCD08068111
  • Cas Number : 881445-78-9
  • Molecular Weight : 242.2732

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9010748

Safety Information

MSDS https://www.hit2lead.com/sds/9010748_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart