CH4072933914 ChemBridge Corporation

3-(4-aminophenyl)-N-cyclopropylpropanamide

  • MDL number MFCD05857485
  • Cas Number : 698992-37-9
  • Molecular Weight : 204.2682

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9038592

Safety Information

MSDS https://www.hit2lead.com/sds/9038592_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart