CH4072934700 ChemBridge Corporation

3-amino-N-cyclohexyl-N-methylbenzamide

  • MDL number MFCD02182182
  • Cas Number : 675112-76-2
  • Molecular Weight : 232.3214

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9040379

Safety Information

MSDS https://www.hit2lead.com/sds/9040379_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart