CH4072937472 ChemBridge Corporation

4-(2,3-dihydro-1H-inden-5-ylamino)-4-oxobutanoic acid

  • MDL number MFCD06065713
  • Cas Number : 847588-85-6
  • Molecular Weight : 233.2631

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9046110

Safety Information

MSDS https://www.hit2lead.com/sds/9046110_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart