CH4100617938 ChemBridge Corporation

N-(3-aminophenyl)-3-phenylpropanamide

  • MDL number MFCD08691856
  • Cas Number : 754162-13-5
  • Molecular Weight : 240.3003

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9034905

Safety Information

MSDS https://www.hit2lead.com/building-blocks/9034905
Availability details for
Quantity
  • Availability
  • Add to Cart