CH4100617990 ChemBridge Corporation

2-[(4-chlorobenzyl)thio]propanoic acid

  • MDL number MFCD04070622
  • Cas Number : 122305-66-2
  • Molecular Weight : 230.7111

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9012800

Safety Information

MSDS https://www.hit2lead.com/sds/9012800_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart