CH4100874334 ChemBridge Corporation

2-(3-acetyl-1H-indol-1-yl)butanoic acid

  • MDL number MFCD08691809
  • Cas Number : 869949-98-4
  • Molecular Weight : 245.2738

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7992369

Safety Information

MSDS https://www.hit2lead.com/sds/7992369_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart