CH4101217801 ChemBridge Corporation

N-(3-hydroxyphenyl)-2-furamide

  • MDL number MFCD00626273
  • Cas Number : 55679-22-6
  • Molecular Weight : 203.1941

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7731776

Safety Information

MSDS https://www.hit2lead.com/sds/7731776_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart