CH4101217937 ChemBridge Corporation

2-amino-N-methyl-N-phenylbenzamide

  • MDL number MFCD00443942
  • Cas Number : 6632-37-7
  • Molecular Weight : 226.2738

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7887216

Safety Information

MSDS https://www.hit2lead.com/sds/7887216_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart