CH4102022153 ChemBridge Corporation

3-{[5-(4-pyridinyl)-1,3,4-oxadiazol-2-yl]thio}propanoic acid

  • MDL number MFCD03825234
  • Cas Number : 604740-20-7
  • Molecular Weight : 251.2618

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7928684

Safety Information

MSDS https://www.hit2lead.com/sds/7928684_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart