CH4102149256 ChemBridge Corporation

(2-cyclopropyl-1H-benzimidazol-1-yl)acetic acid

  • MDL number MFCD08691428
  • Cas Number : 97968-85-9
  • Molecular Weight : 216.2359

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4002576

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4002576
Availability details for
Quantity
  • Availability
  • Add to Cart