CH4102149298 ChemBridge Corporation

2-(5-methyl-2-thienyl)ethanamine

  • MDL number MFCD08059759
  • Cas Number : 30433-92-2
  • Molecular Weight : 141.2339

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4004130

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4004130
Availability details for
Quantity
  • Availability
  • Add to Cart