CH4102149442 ChemBridge Corporation

1-benzothiophene-2-sulfonyl chloride

  • MDL number MFCD03659701
  • Cas Number : 90001-64-2
  • Molecular Weight : 232.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4002793

Safety Information

signalword Danger
hcodes 8
MSDS https://www.hit2lead.com/sds/4002793_sds.pdf
hstatements H314, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart