CH4102149509 ChemBridge Corporation

5-isobutylisoxazole-3-carboxylic acid

  • MDL number MFCD04113974
  • Cas Number : 150517-80-9
  • Molecular Weight : 169.1778

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003154

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4003154
Availability details for
Quantity
  • Availability
  • Add to Cart