CH4102149512 ChemBridge Corporation

(4-phenylpiperidin-1-yl)acetic acid

  • MDL number MFCD04038699
  • Cas Number : 799266-57-2
  • Molecular Weight : 219.2796

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003158

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4003158
Availability details for
Quantity
  • Availability
  • Add to Cart