CH4102149581 ChemBridge Corporation

2-chloro-4,5-dimethoxybenzaldehyde

  • MDL number MFCD00461433
  • Cas Number : 18093-05-5
  • Molecular Weight : 200.619

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003462

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4003462
Availability details for
Quantity
  • Availability
  • Add to Cart