CH4102149582 ChemBridge Corporation

4-azepan-1-ylbutan-1-ol

  • MDL number MFCD03211285
  • Cas Number : 114960-98-4
  • Molecular Weight : 171.2798

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4001155

Safety Information

MSDS https://www.hit2lead.com/sds/4001155_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart