CH4102149678 ChemBridge Corporation

2,1,3-benzoxadiazole-5-carbaldehyde

  • MDL number MFCD01200350
  • Cas Number : 32863-33-5
  • Molecular Weight : 148.1189

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003957

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4003957
Availability details for
Quantity
  • Availability
  • Add to Cart