CH4102149705 ChemBridge Corporation

(1-methyl-2-oxo-2,3-dihydro-1H-indol-3-yl)acetic acid

  • MDL number MFCD07186396
  • Cas Number : 21591-75-3
  • Molecular Weight : 205.2099

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4004159

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4004159
Availability details for
Quantity
  • Availability
  • Add to Cart