CH4102149752 ChemBridge Corporation

N-methyl-1-[4-(trifluoromethyl)phenyl]methanamine

  • MDL number MFCD04115411
  • Cas Number : 90390-11-7
  • Molecular Weight : 189.1776

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4004319

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4004319
Availability details for
Quantity
  • Availability
  • Add to Cart